Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Educational Insights GeoSafari™ Jr. My First Microscope
SCI_ED

Catalog No. S02734
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02734 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02734 Supplier Educational Insights Supplier No. 5112
Add to Cart
Add to Cart

GeoSafari Jr. My First Microscope allows preschoolers to see magnified hidden worlds. This fully functional microscope is the perfect introduction to a real science tool and STEM learning. My First Microscope has a range of magnification (2.5x to 8x) designed for toddlers and preschoolers.

  • GeoSafari Jr. My First Microscope allows preschoolers to see magnified hidden worlds with ease
  • This fully functional microscope is the perfect introduction to a real science tool and STEM learning.
  • My First Microscope has a range of magnification (2.5x to 8x) designed for toddlers and preschoolers.
  • Magnified views can be challenging for little ones to understand, so by starting with the 2.5x view (with the focusing knob turned as far away from your child as it will go), your preschooler can orient themself to the item they're viewing, and as they turn the focusing knob towards themself, they can gradually pull away to an 8x view.
  • The wide field of view of these lower magnifications is critical to little ones' understanding.
  • My First Microscope also features two extra-large eyepieces (no need to close one eye!) and a chunky, kid-sized focusing knob.
  • A bright LED with easy push-button action lights up details for even more discoveries.
  • Requires 3 AAA batteries (not included).

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type GeoSafari Toy Microscope
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.