Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Educational Insights GeoSafari Motorized Solar System
SCI_ED

Catalog No. S02728
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02728 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02728 Supplier Educational Insights Supplier No. 5287

missing translation for 'validMainframeMsg'

Bring the magic of space home with the GeoSafari Motorized Solar System! Eight colorful planets rotate around a glowing sun, just like they do in space! Remove the star dome cover to project the constellations on the wall or ceiling. Requires 4 AA batteries (not included).

  • Bring the magic of space home with the GeoSafari Motorized Solar System
  • Eight colorful planets rotate around a glowing sun, just like they do in space
  • Remove the star dome cover to project the constellations on the wall or ceiling.
  • Requires 4 AA batteries (not included).

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Class
Product Type GeoSafari Solar System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.