Learn More
FUJIFILM Biosciences Geosmin Standard
Catalog No. 5000450362
Encompass_Preferred
Geosmin is one source of the earthy-musty taste in water Early detection of Geosmin in source water is helpful in preventing an odor problem These problems have been reported when levels are as little as 10-15 parts per trillion Geosmin Standard is synthetically produced and is a mixture of the D- and L- forms of trans-110-dimethyl-trans-9decanol This standard enables precise quantification using capillary gas chromatography With high purity and reliable stability it serves as an essential standard for researchers and environmental analysts concerned with water quality
Catalog No. 50-004-50362 Supplier FUJIFILM Biosciences Supplier No. 07701911
Geosmin is one source of the earthy-musty taste in water Early detection of Geosmin in source water is helpful in preventing an odor problem These problems have been reported when levels are as little as 10-15 parts per trillion Geosmin Standard is synthetically produced and is a mixture of the D- and L- forms of trans-110-dimethyl-trans-9decanol This standard enables precise quantification using capillary gas chromatography With high purity and reliable stability it serves as an essential standard for researchers and environmental analysts concerned with water quality