Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gepirone-d8 | 2749331-28-8 | 98.0% | 5 MG
SDP

Catalog No. 5000230712
Encompass_Preferred

Gepirone-d8 is the deuterium-labeled isotopologue of gepirone supplied as a stable-isotope reference standard for analytical and pharmacokinetic studies.

  • Deuterium-labeled internal standard for gepirone.
  • Intended for use in mass spectrometry and pharmacokinetic studies.
  • Molecular formula C19H21D8N5O2, molecular weight 367.52 g/mol.
  • Purity 98.0% (reported).
  • CAS number 2749331-28-8.
  • Soluble in DMSO; provided as a 5 mg solid.

Catalog No. 50-002-30712 Supplier Medchemexpress LLC Supplier No. HY122422S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gepirone-d8 is the deuterium-labeled isotopologue of gepirone supplied as a stable-isotope reference standard for analytical and pharmacokinetic studies.

  • Deuterium-labeled internal standard for gepirone.
  • Intended for use in mass spectrometry and pharmacokinetic studies.
  • Molecular formula C19H21D8N5O2, molecular weight 367.52 g/mol.
  • Purity 98.0% (reported).
  • CAS number 2749331-28-8.
  • Soluble in DMSO; provided as a 5 mg solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.