Learn More
Medchemexpress LLC Gepotidacin | 1075236-89-3 | 99.9% | 448.52 | 5 MG
SDP
Catalog No. 5000395462
Encompass_Preferred
Gepotidacin is a novel triazaacenaphthylene bacterial type II topoisomerase inhibitor. It is a first-in-class antibacterial that inhibits bacterial DNA gyrase and topoisomerase IV through a unique mechanism. It demonstrates in vitro activity against a broad spectrum of gram-negative and gram-positive bacteria, including drug-resistant strains.
  • Novel triazaacenaphthylene bacterial type II topoisomerase inhibitor
  • Inhibits bacterial DNA gyrase and topoisomerase IV
  • Effective against gram-negative and gram-positive bacteria
  • Active against drug-resistant bacterial strains
  • Targets pathogens associated with conventional and biothreat infections
Catalog No. 50-003-95462 Supplier Medchemexpress LLC Supplier No. HY167425MG
Gepotidacin is a novel triazaacenaphthylene bacterial type II topoisomerase inhibitor. It is a first-in-class antibacterial that inhibits bacterial DNA gyrase and topoisomerase IV through a unique mechanism. It demonstrates in vitro activity against a broad spectrum of gram-negative and gram-positive bacteria, including drug-resistant strains.
  • Novel triazaacenaphthylene bacterial type II topoisomerase inhibitor
  • Inhibits bacterial DNA gyrase and topoisomerase IV
  • Effective against gram-negative and gram-positive bacteria
  • Active against drug-resistant bacterial strains
  • Targets pathogens associated with conventional and biothreat infections