Learn More
Medchemexpress LLC HY-N0472 5mg Medchemexpress, Geraniin CAS:60976-49-0 Purity:>98%
SDP
Catalog No. 501933587
Encompass_Preferred
Medchemexpress, HY-N0472 5mg Geraniin CAS:60976-49-0 Geraniin is a TNF-α releasing inhibitor with numerous activities including anticancer, anti-inflammatory, and anti-hyperglycemic activities, with an IC 50 of 43 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3587 Supplier Medchemexpress LLC Supplier No. HYN04725MG
Medchemexpress, HY-N0472 5mg Geraniin CAS:60976-49-0 Geraniin is a TNF-α releasing inhibitor with numerous activities including anticancer, anti-inflammatory, and anti-hyperglycemic activities, with an IC 50 of 43 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.