Learn More
Medchemexpress LLC HY-N6952 100mg Medchemexpress, Geraniol CAS:106-24-1 Purity:>98%
SDP
Catalog No. 501970483
Encompass_Preferred
Medchemexpress, HY-N6952 100mg Geraniol CAS:106-24-1 Geraniol, an olefinic terpene, was found to inhibit growth of Candida albicans and Saccharomyces cerevisiae strains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0483 Supplier Medchemexpress LLC Supplier No. HYN6952100MG
Medchemexpress, HY-N6952 100mg Geraniol CAS:106-24-1 Geraniol, an olefinic terpene, was found to inhibit growth of Candida albicans and Saccharomyces cerevisiae strains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.