Learn More
Medchemexpress LLC Geraniol-d6 (major) | 1660976-93-1 | 95.3% | 160.29 g/mol | C10H12D6O | 500ug
SDP
Catalog No. 5000269446
Encompass_Preferred
Geraniol-d6 (Major) is the deuterated labeled Geraniol (Major) Geraniol is an olefin terpene with oral activity Geraniol inhibits cell proliferation and promotes apoptosis Geraniol has antibacterial antifungal antioxidant anti-inflammatory and antitumor activities Geraniol can be used to study diabetes[1][2][3][4][5]
Catalog No. 50-002-69446 Supplier Medchemexpress LLC Supplier No. HYW782079500UG
Geraniol-d6 (Major) is the deuterated labeled Geraniol (Major) Geraniol is an olefin terpene with oral activity Geraniol inhibits cell proliferation and promotes apoptosis Geraniol has antibacterial antifungal antioxidant anti-inflammatory and antitumor activities Geraniol can be used to study diabetes[1][2][3][4][5]