Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Geraniol (Standard) | 106-24-1 | 25 MG
SDP

Catalog No. 5000420548
Encompass_Preferred

Geraniol (Standard) is an analytical reference standard. It is an olefinic terpene that inhibits the growth of Candida albicans and Saccharomyces cerevisiae strains. This product is intended for research and analytical applications, serving as a reference for qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analytical applications
  • Used as a reference in HPLC, GC, and MS experiments
  • Inhibits Candida albicans and Saccharomyces cerevisiae strains
  • Molecular weight of 154.25
  • Chemical formula C₁₀H₁₈O
  • Colorless to light yellow liquid appearance
  • Purity of 96.88%
  • Store at 4°C, sealed, away from light and moisture

Catalog No. 50-004-20548 Supplier Medchemexpress LLC Supplier No. 25MGHYN6952R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Geraniol (Standard) is an analytical reference standard. It is an olefinic terpene that inhibits the growth of Candida albicans and Saccharomyces cerevisiae strains. This product is intended for research and analytical applications, serving as a reference for qualitative, quantitative, and methodological research experiments.

  • Analytical standard for research and analytical applications
  • Used as a reference in HPLC, GC, and MS experiments
  • Inhibits Candida albicans and Saccharomyces cerevisiae strains
  • Molecular weight of 154.25
  • Chemical formula C₁₀H₁₈O
  • Colorless to light yellow liquid appearance
  • Purity of 96.88%
  • Store at 4°C, sealed, away from light and moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.