Learn More
Apexbio Technology LLC GERANYL-ACETATE-100MG
SDP
Catalog No. 5000568585
Encompass_Preferred
Geranyl acetate (CAS No 105-87-3) is a monoterpene ester commonly explored in biomedical research due to its natural occurrence in essential oils of plants like lemongrass and sweet marjoram It is primarily studied for its potential role in modulating cellular pathways involved in anti-inflammatory and anticancer activities Geranyl acetate exhibits in vitro activity in the nanomolar to low micromolar range which indicates its potency in biological systems Typical applications include cell-based assays and animal studies for drug screening and understanding its pharmacodynamics Experimental concentrations typically depend on the specific research design Researchers leverage its capability to impact signaling pathways like NF- B and MAPK which are crucial in inflammation and cancer development These studies aim to uncover therapeutic potentials and safety profiles contributing to developing novel treatments for various conditions
Catalog No. 50-005-68585 Supplier Apexbio Technology LLC Supplier No. C6502100
Geranyl acetate (CAS No 105-87-3) is a monoterpene ester commonly explored in biomedical research due to its natural occurrence in essential oils of plants like lemongrass and sweet marjoram It is primarily studied for its potential role in modulating cellular pathways involved in anti-inflammatory and anticancer activities Geranyl acetate exhibits in vitro activity in the nanomolar to low micromolar range which indicates its potency in biological systems Typical applications include cell-based assays and animal studies for drug screening and understanding its pharmacodynamics Experimental concentrations typically depend on the specific research design Researchers leverage its capability to impact signaling pathways like NF- B and MAPK which are crucial in inflammation and cancer development These studies aim to uncover therapeutic potentials and safety profiles contributing to developing novel treatments for various conditions