Learn More
Medchemexpress LLC Geranyl acetate | 105-87-3 | 1 ML
SDP
Catalog No. 5000418953
Encompass_Preferred
Geranyl acetate is an acyclic monoterpene ester derived from geraniol. Widely used in the cosmetics industry for its pleasant scent, it also acts as an apoptosis activator, showing significant anti-cancer activity against colo-205 cancer cells.
  • Induces cell apoptosis
  • Exhibits significant anti-cancer activity against colo-205 cancer cells
  • Apoptosis associated with upregulation of Bax and downregulation of Bcl-2 expressions
  • Acyclic monoterpene ester derived from geraniol
  • Widely used in the cosmetics industry for its pleasant scent
  • High purity of 99.74%
  • Appearance: colorless to light yellow liquid
Catalog No. 50-004-18953 Supplier Medchemexpress LLC Supplier No. HYN707010MM/1ML
Geranyl acetate is an acyclic monoterpene ester derived from geraniol. Widely used in the cosmetics industry for its pleasant scent, it also acts as an apoptosis activator, showing significant anti-cancer activity against colo-205 cancer cells.
  • Induces cell apoptosis
  • Exhibits significant anti-cancer activity against colo-205 cancer cells
  • Apoptosis associated with upregulation of Bax and downregulation of Bcl-2 expressions
  • Acyclic monoterpene ester derived from geraniol
  • Widely used in the cosmetics industry for its pleasant scent
  • High purity of 99.74%
  • Appearance: colorless to light yellow liquid