Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Geranyl acetate | 105-87-3 | 100 G
SDP

Catalog No. 5000419641
Encompass_Preferred

Geranyl acetate is an acyclic monoterpene ester derived from geraniol. It is widely used in the cosmetics industry due to its pleasant scent and has been shown to induce cell apoptosis. This product appears as a colorless to light yellow liquid.

  • Acyclic monoterpene ester
  • Derived from geraniol
  • Used in cosmetics industry for pleasant scent
  • Induces cell apoptosis
  • Purity: 99.74%
  • Molecular weight: 196.29
  • Chemical formula: C12H20O2
  • Density: 0.916 g/cm³
  • Exhibits significant anti-cancer activity against colo-205 cancer cell line
  • Agonist activity at rat TRPA1 (EC50 of 25.8 μM in HEK293 cells)
  • Antagonist activity against rat TRPA1 (IC50 of 21.2 μM in HEK293 cells)

Catalog No. 50-004-19641 Supplier Medchemexpress LLC Supplier No. HYN7070100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Geranyl acetate is an acyclic monoterpene ester derived from geraniol. It is widely used in the cosmetics industry due to its pleasant scent and has been shown to induce cell apoptosis. This product appears as a colorless to light yellow liquid.

  • Acyclic monoterpene ester
  • Derived from geraniol
  • Used in cosmetics industry for pleasant scent
  • Induces cell apoptosis
  • Purity: 99.74%
  • Molecular weight: 196.29
  • Chemical formula: C12H20O2
  • Density: 0.916 g/cm³
  • Exhibits significant anti-cancer activity against colo-205 cancer cell line
  • Agonist activity at rat TRPA1 (EC50 of 25.8 μM in HEK293 cells)
  • Antagonist activity against rat TRPA1 (IC50 of 21.2 μM in HEK293 cells)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.