Learn More
Selleck Chemical LLC Geranyl acetate S5091-25ul
SDP
Catalog No. 503110929
Encompass_Preferred
Geranyl acetate (Geraniol acetate Geranyl ethanoate) is a natural organic compound that is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient Geranyl acetate exhibits anti-cancer effects by inducing cell apoptosis
Catalog No. 50-311-0929 Supplier Selleck Chemical LLC Supplier No. S509125UL
Geranyl acetate (Geraniol acetate Geranyl ethanoate) is a natural organic compound that is used primarily as a component of perfumes for creams and soaps and as a flavoring ingredient Geranyl acetate exhibits anti-cancer effects by inducing cell apoptosis