Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Geranyl acetate

Catalog No. 173495100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-349-5100G 100 g
17-349-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-349-5100G Supplier Sigma Aldrich Supplier No. 173495100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Geranyl acetate is an aroma active rice volatile
  • It is one of the components of the essential oils of daucus carota L
  • Seeds

Specifications

CAS 105-87-3
Density 0.916 g/mL (at 25°C)
Boiling Point 138°C (25 mmHg)
Molecular Formula C12H20O2
Refractive Index n20/D 1.461 (literature)
Linear Formula CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
MDL Number MFCD00015037
Quantity 100 g
Synonym trans-3,7-Dimethyl-2,6-octadien-1-yl acetate; trans-3,7-Dimethyl-2,6-octadienyl acetate
Molecular Weight (g/mol) 196.29
Percent Purity ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.