Learn More
Sigma Aldrich Fine Chemicals Biosciences Geranyl butyrate natural 91KG
Catalog No. 502908006
Encompass
Geranyl butyrate is one of the key esters that may be contributing to the characteristic flavor of jamun fruit. It also occurs in tomato juice and tomato-based dried products.
Catalog No. 50-290-8006 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2512161KGK
Geranyl butyrate is one of the key esters that may be contributing to the characteristic flavor of jamun fruit. It also occurs in tomato juice and tomato-based dried products.