Learn More
Echelon Biosciences Research Labs GERANYL DIPHOSPHATE 1 MG
SDP
Supplier:  Echelon Biosciences Research Labs I01001MG
Encompass
Geranyl Diphosphate (tris-ammonium salt), Molecular Formula: - C10H29N3O7P2 Molecular Weight (g/mol): -365.30 Storage: -0 deg.C, or flash-frozen with liquid N2 and kept at -70 deg.C. This item is non returnable
Catalog No. 50-720-383