Learn More
Medchemexpress LLC Geranyl diphosphate 1mg | 116057-55-7 | 1 MG
SDP
Catalog No. 5000229535
Encompass_Preferred
Geranyl diphosphate triammonium is the triammonium salt of geranyl pyrophosphate, an isoprenoid intermediate used in terpene and sterol biosynthesis. Supplied as a high-purity reagent for research applications, it is suitable for enzymatic and biochemical studies.
  • High purity suitable for biochemical assays.
  • Available in small milligram pack sizes for research use.
  • Water-soluble triammonium form for aqueous assay compatibility.
  • Used as a substrate or intermediate in terpene biosynthesis studies.
Catalog No. 50-002-29535 Supplier Medchemexpress LLC Supplier No. HY114295A1MG
Geranyl diphosphate triammonium is the triammonium salt of geranyl pyrophosphate, an isoprenoid intermediate used in terpene and sterol biosynthesis. Supplied as a high-purity reagent for research applications, it is suitable for enzymatic and biochemical studies.
  • High purity suitable for biochemical assays.
  • Available in small milligram pack sizes for research use.
  • Water-soluble triammonium form for aqueous assay compatibility.
  • Used as a substrate or intermediate in terpene biosynthesis studies.