Learn More
Medchemexpress LLC Geranyl diphosphate 500ug | 500UG
SDP
Catalog No. 5000229108
Encompass_Preferred
Geranyl diphosphate triammonium is a key intermediate in the isoprenoid biosynthesis pathway (IBP) Geranyl diphosphate triammonium plays key roles in cellular metabolism and is responsible for the production of both sterol and non-sterol isoprenoids[1][2]
Catalog No. 50-002-29108 Supplier Medchemexpress LLC Supplier No. HY114295A500UG
Geranyl diphosphate triammonium is a key intermediate in the isoprenoid biosynthesis pathway (IBP) Geranyl diphosphate triammonium plays key roles in cellular metabolism and is responsible for the production of both sterol and non-sterol isoprenoids[1][2]