Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geranyl linalool, ca. 95%, Tech., mixture of isomers

Catalog No. AC278360250
Change view
Click to view available options
Quantity:
2.5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC278360250 25 g
AC278360025 2.5 g
AC278360500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC278360250 Supplier Thermo Scientific Chemicals Supplier No. 278360250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1113-21-9 | C20H34O | 290.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-21-9
Molecular Formula C20H34O
Molecular Weight (g/mol) 290.49
MDL Number MFCD00059363
InChI Key IQDXAJNQKSIPGB-HQSZAHFGNA-N
Synonym geranyl linalool, geranyllinalool, 3,7,11,15-tetramethyl-1,6,10,14-hexadecatetraen-3-ol, e,e-geranyllinalool, 6e,10e-geranyllinalool, 3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, 6e,10e-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, e,e-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol, 1,6,10,14-hexadecatetraen-3-ol, 3,7,11,15-tetramethyl-, 6e,10e, 1,6,10,14-hexadecatetraen-3-ol, 3,7,11,15-tetramethyl
PubChem CID 5365872
ChEBI CHEBI:74299
IUPAC Name (6E,10E)-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CCC(C)(O)C=C

Specifications

Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 250.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4920
Quantity 25 g
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents, solubel in 70% ethanol
Formula Weight 290.48
Percent Purity ca. 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Geranyl linalool, tech., ca. 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 214-201-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.