Learn More
Echelon Biosciences Research Labs GERANYL S-THIOLODIPHOSPHATE 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs IS1001MG
Encompass
Geranyl thiolodiphosphate (GSPP) is an analog of GPP in which the non-bridging oxygen has been replaced by sulfur resulting in a less-reactive compound. GSPP is turned over by avian farnesyl diphosphate synthase (FPPase) more slowly than FPP and acts as a competitive inhibitor (KI = 24.8 M). This item is non returnable
Catalog No. 50-720-412