Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Geranylamine

Catalog No. 4126435G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-264-35G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-264-35G Supplier Sigma Aldrich Supplier No. 4126435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
  • Sigma-aldrich does not collect analytical data for this product
  • Buyer assumes responsibility to confirm product identity and/or purity
  • All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise

Specifications

CAS 6246-48-6
Density 0.829 g/mL (at 25°C (literature))
Boiling Point 105°C (19 mmHg)
Molecular Formula C10H19N
Refractive Index n20/D 1.476 (literature)
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)=CHCH2NH2
Quantity 1 g
MDL Number MFCD00192193
Synonym trans-3,7-Dimethyl-2,6-octadien-1-ylamine; trans-3,7-Dimethyl-2,6-octadienyl amine
Molecular Weight (g/mol) 153.26
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.