Learn More
Echelon Biosciences Research Labs GERANYLGERANIOL 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs S02001MG
Encompass
(E,E,E)-Geranylgeraniol is an intermediate in the biosynthetic pathway of vitamins E & K and is linked to proteins via post-translational modification. This item is non returnable
Catalog No. 50-720-754