Learn More
Echelon Biosciences Research Labs GERANYLGERANIOL 10 mg
SDP
Catalog No. 50720755
Encompass
(E,E,E)-Geranylgeraniol is an intermediate in the biosynthetic pathway of vitamins E & K and is linked to proteins via post-translational modification. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-755 Supplier Echelon Biosciences Research Labs Supplier No. S020010MG
(E,E,E)-Geranylgeraniol is an intermediate in the biosynthetic pathway of vitamins E & K and is linked to proteins via post-translational modification. This item is non returnable