Learn More
Enzo Life Sciences Geranylgeraniol (20mg). CAS: 24034-73-9
SDP
Supplier:  Enzo Life Sciences BMLG2430020
Encompass_Preferred
Precursor to geranylgeranylpyrophosphate. Induces apoptosis in HL-60 cells. Purity: ≥95% (TLC). Solubility: Soluble in chloroform or ethyl acetate. Long Term Storage: -80°C.
Catalog No. 50-201-0873