Learn More
Medchemexpress LLC Geranylgeraniol | 24034-73-9 | 99.2% | 290.48 | 200 MG
SDP
Catalog No. 5000455142
Encompass_Preferred
Geranylgeraniol is an orally active subtype of vitamin K2 and an intermediate of the mevalonate pathway. It targets the NF-κB signaling pathway and has shown potential to alleviate LPS-induced microglial inflammation in animal models.
  • Targets NF-κB signaling pathway
  • Alleviates LPS-induced microglial inflammation in animal models
  • For research use only
  • Ships at room temperature in continental US
  • Can be stored at -20°C for 3 years in pure form
  • Can be stored at -80°C for 6 months in solvent
Catalog No. 50-004-55142 Supplier Medchemexpress LLC Supplier No. HYW011474200MG
Geranylgeraniol is an orally active subtype of vitamin K2 and an intermediate of the mevalonate pathway. It targets the NF-κB signaling pathway and has shown potential to alleviate LPS-induced microglial inflammation in animal models.
  • Targets NF-κB signaling pathway
  • Alleviates LPS-induced microglial inflammation in animal models
  • For research use only
  • Ships at room temperature in continental US
  • Can be stored at -20°C for 3 years in pure form
  • Can be stored at -80°C for 6 months in solvent