Learn More
Weber Scientific GERBER BOTTLE BULB BRUSH
SDP
Supplier:  Weber Scientific 102900
Encompass
Specially designed for cleaning the large bulb of the Gerber bottle. Heavy duty! (Not for 4% bottles.)
Catalog No. NC2260196