Learn More
MSC PortionPac Germicidal Cleaner, Packet, 6.5 oz, EPA Registered, 96 Pieces
Catalog No. 502868242
Encompass
PortionPac Germicidal Cleaner is an EPA registered solution efficacy-tested as an effective bactericide, fungicide, virucide, and mildewcide. It effectively functions in hard water (up to 400 ppm) and in the presence of organic soil, suitable for cleaning floors, walls, toilets, tables, mirrors, and appliance exteriors.
  • Efficacy against bacteria, fungi, viruses, and mildew
  • Effective in hard water up to 400 ppm and presence of organic soil
  • Versatile for use on multiple hard surfaces
  • Green Seal certified and biodegradable
  • Made from recycled content
  • Each pouch yields 7 gal. of solution
  • Causes skin irritation and serious eye damage (GHS Classification)
Catalog No. 50-286-8242 Supplier MSC Supplier No. 68116995
PortionPac Germicidal Cleaner is an EPA registered solution efficacy-tested as an effective bactericide, fungicide, virucide, and mildewcide. It effectively functions in hard water (up to 400 ppm) and in the presence of organic soil, suitable for cleaning floors, walls, toilets, tables, mirrors, and appliance exteriors.
  • Efficacy against bacteria, fungi, viruses, and mildew
  • Effective in hard water up to 400 ppm and presence of organic soil
  • Versatile for use on multiple hard surfaces
  • Green Seal certified and biodegradable
  • Made from recycled content
  • Each pouch yields 7 gal. of solution
  • Causes skin irritation and serious eye damage (GHS Classification)