Learn More
Medchemexpress LLC Germacrene D | 23986-74-5 | >97.0% | 204.35 g·mol⁻¹ | C15H24 | 1MG
SDP
Catalog No. 5000239624
Encompass_Preferred
Germacrene D is a naturally occurring sesquiterpene (C15H24, MW 204.35 g·mol⁻¹) isolated from Solidago canadensis and provided as a research-grade reference compound. It is reported to have antibacterial and antifungal activity and has been used as an adjuvant with aminoglycoside antibiotics and azole antifungals.
  • Natural sesquiterpene isolated from Solidago canadensis.
  • Reported antibacterial and antifungal activity.
  • Acts as an adjuvant with aminoglycosides and azoles.
  • High purity (≥97.0% per product specification; batch-specific values may vary).
  • Molecular formula C15H24; molecular weight 204.35 g·mol⁻¹.
  • Available in micro-scale pack sizes suitable for analytical and biological assays.
Catalog No. 50-002-39624 Supplier Medchemexpress LLC Supplier No. HY1256851MG
Germacrene D is a naturally occurring sesquiterpene (C15H24, MW 204.35 g·mol⁻¹) isolated from Solidago canadensis and provided as a research-grade reference compound. It is reported to have antibacterial and antifungal activity and has been used as an adjuvant with aminoglycoside antibiotics and azole antifungals.
  • Natural sesquiterpene isolated from Solidago canadensis.
  • Reported antibacterial and antifungal activity.
  • Acts as an adjuvant with aminoglycosides and azoles.
  • High purity (≥97.0% per product specification; batch-specific values may vary).
  • Molecular formula C15H24; molecular weight 204.35 g·mol⁻¹.
  • Available in micro-scale pack sizes suitable for analytical and biological assays.