Learn More
Medchemexpress LLC Germacrone Standard 1mg
SDP
Catalog No. 5000669504
Encompass_Preferred
Germacrone (Standard) is an analytical standard of Gemmacrone (HY-N0440) This product is intended for research and analytical applications Germacrone (Standard) is a sesquiterpene compound with multiple biological activities Germacrone (Standard) inhibits the H1N1 and H3N2 in uenza A viruses and the in uenza B virus Germacrone (Standard) blocks the progressionof arthritis by regulating Th1/Th2 balance and inhibiting NF- B signaling Germacrone (Standard) can arrest the cell cycle at G0/G1 and G2/M phases and induce apoptosis in breast cancer cells Germacrone (Standard) inhibits 5 -reductase and has anti-androgenic effect Germacrone (Standard) has neuroprotective functions and can be used for the study of traumatic brain injury (TBI) Germacrone (Standard) also has antioxidant activity[1][2][3][4][5]
Catalog No. 50-006-69504 Supplier Medchemexpress LLC Supplier No. 1MGHYN0440R
Germacrone (Standard) is an analytical standard of Gemmacrone (HY-N0440) This product is intended for research and analytical applications Germacrone (Standard) is a sesquiterpene compound with multiple biological activities Germacrone (Standard) inhibits the H1N1 and H3N2 in uenza A viruses and the in uenza B virus Germacrone (Standard) blocks the progressionof arthritis by regulating Th1/Th2 balance and inhibiting NF- B signaling Germacrone (Standard) can arrest the cell cycle at G0/G1 and G2/M phases and induce apoptosis in breast cancer cells Germacrone (Standard) inhibits 5 -reductase and has anti-androgenic effect Germacrone (Standard) has neuroprotective functions and can be used for the study of traumatic brain injury (TBI) Germacrone (Standard) also has antioxidant activity[1][2][3][4][5]