Learn More
Germaine™ Laboratories AimStrip ™ Plus Blood Glucose Testing System Accessories: Controls Solution
Catalog No. 23111278
Change view
Click to view available options
Quantity:
2
Catalog No. Quantity
23-111-278 2
1 options
This item is not returnable. View return policy
Catalog No. 23-111-278 Supplier Germaine™ Laboratories Supplier No. 37312
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Additional controls solutions for AimStrip Plus Blood Glucose Testing System
For use with AimStrip Plus Blood Glucose Testing System (23-111-275).
Specifications
Product Type AimStrip™ plus Glucose Control Solution
Sample Size 2 mL
For Use With (Application) Used for testing blood glucose levels
For Use With (Equipment) AimStrip™ plus Blood Glucose meter
Quantity 2