Learn More
Germaine™ Laboratories AimStrip ™ Plus Blood Glucose Testing System Accessories: Test Strips
Catalog No. 23111276
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
23-111-276 50
1 options
This item is not returnable. View return policy
Catalog No. 23-111-276 Supplier Germaine™ Laboratories Supplier No. 37350
Add to Cart
Add to Cart
This item is not returnable. View return policy
AimStrip Plus BG Test Strips are for the quantitative measurement of glucose in Capillary whole blood
For use with AimStrip Plus Blood Glucose Testing System (23-111-275).
Specifications
Product Type AimStrip™ plus Blood Glucose Test Strip
Analysis Time 10 sec.
Sample Size 1 μL
For Use With (Application) Used for testing blood glucose levels
For Use With (Equipment) AimStrip™ plus Blood Glucose meter
Quantity 50