Learn More
Germaine™ Laboratories AimStrip ™ Plus Blood Glucose Testing System
Catalog No. 23111275
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-111-275 Each
1 options
Catalog No. 23-111-275 Supplier Germaine™ Laboratories Supplier No. 37321
Add to Cart
Add to Cart
Includes one meter, carrying case, 10 lancets, lancing device, logbook, and user manual
  • Only 1μL blood sample required
  • Results in 10 seconds
  • Code chip lasts the life of the meter
  • Low-cost glucose strips
  • Stores up to 300 tests with date and time
Order Info
Additional test strips (23-111-276), lancets (23-111-277) and controls solution (23-111-278) available separately.
Specifications
Product Type AimStrip™ plus Blood Glucose Meter Kit
Analysis Time 10 sec.
Sample Size 1 μL
For Use With (Application) Use with AimStrip™ plus Blood Glucose test strips for testing Blood Glucose levels
Includes 1 Meter, carrying case, 10 lancets, lancing device, logbook, user's manuals English and Spanish, warranty card
Quantity Each