Learn More
Germaine™ Laboratories AimStrip™ Urine Auto Analyzer
Description
- CLIA waived
- High through put, up to 500 tests per hour
- Large touch screen for easy navigation
- Automatic calibration for accurate results
- Automatically saves the last 2000 results
- Reads 4, 8, and 10 parameter strips
- Test modes include routine, stat and QC
- Includes RS232C and USB ports to export results
- Program languages: English, Spanish and Portuguese
Specifications
Specifications
| Product Type | Urine Auto Analyzer |
| Display Type | Touch Screen |
| Languages | English, Spanish, Portuguese |
| Memory | 2000 results |
| For Use With (Application) | Semi-quantitative determination of glucose, bilirubin, ketone, specific gravity, blood, pH, protein, urobilinogen, leukocytes and qualitative detection of nitrite in urine |