Learn More
Germaine™ Laboratories Febrile Antigens For Proteus OX
Catalog No. 16414105
Change view
Click to view available options
Type:
Proteus OX19 Antigen
Proteus OX2 Antigen
Proteus OXK Antigen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
16-414-105 Proteus OXK Antigen
16-414-106 Proteus OX19 Antigen
16-414-104 Proteus OX2 Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-414-105 Supplier Germaine™ Laboratories Supplier No. 1200
Add to Cart
Add to Cart
Febrile Antigens intended for detection of bacterial agglutinins associated with Proteus OX infection
  • 5mL vial
Specifications
Detectable Analytes Proteus OXK Antibodies
Type Proteus OXK Antigen
Quantity 5 mL
Professional use only