Learn More
Germaine™ Laboratories Febrile Antigens For Salmonella H
Catalog No. 16414118
Change view
Click to view available options
Type:
Salmonella H a Antigen
Salmonella H b Antigen
Salmonella H d Antigen
Detectable Analytes:
Salmonella H A Antibodies
Salmonella H B Antibodies
Salmonella H D Antibodies
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Detectable Analytes
16-414-118 Salmonella H b Antigen Salmonella H B Antibodies
16-414-117 Salmonella H a Antigen Salmonella H A Antibodies
16-414-120 Salmonella H d Antigen Salmonella H D Antibodies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 16-414-118 Supplier Germaine™ Laboratories Supplier No. 1401
Add to Cart
Add to Cart
This item is not returnable. View return policy
Febrile antigens intended for detection of bacterial agglutinins associated with Salmonella H infection
  • 5mL vial
Specifications
Detectable Analytes Salmonella H B Antibodies
Type Salmonella H b Antigen
Quantity 5 mL
Professional use only