Learn More
Germaine™ Laboratories Febrile Antigens For Salmonella O
Catalog No. 16414101
Change view
Click to view available options
Type:
Salmonella O Group D Antigen
Somatic Antigen
Detectable Analytes:
Salmonella O Group B Agglutinins
Salmonella O Group D Agglutinins
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Detectable Analytes
16-414-101 Somatic Antigen Salmonella O Group B Agglutinins
16-414-103 Salmonella O Group D Antigen Salmonella O Group D Agglutinins
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-414-101 Supplier Germaine™ Laboratories Supplier No. 1501
Add to Cart
Add to Cart
Febrile Antigens intended for detection of bacterial agglutinins associated with Salmonella O infection
  • 5mL vial
Specifications
Detectable Analytes Salmonella O Group B Agglutinins
Type Somatic Antigen
Quantity 5 mL
Professional use only