Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Urinalysis Reagent Test Strips

Catalog No. 23111255
Change view
Click to view available options
Detectable Analytes:
Protein, Glucose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-111-255 Protein, Glucose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-255 Supplier Germaine™ Laboratories Supplier No. 52100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid screening tool detects up to four analytes on one test strip

  • Read strips visually or on instrumentation
  • Rapid qualitative results
  • Shelf life 18 months from date of manufacture
  • CLIA waived

Specifications

Clia Complexity Waived
Detectable Analytes Protein, Glucose
For Use With (Application) Urinalysis
Quantity 100 Tests
Test Time 1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.