Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimTab™ Bilirubin Tablets

Catalog No. 23111288
Change view
Click to view available options
Quantity:
100/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-288 100/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-288 Supplier Germaine™ Laboratories Supplier No. 13200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AimTab™ Bilirubin Tablets are for the detection of bilirubin in urine. Results are read at 60 seconds.

  • For the detection of bilirubin in urine.
  • 100 tablets/bottle
  • Results in 60 seconds!
  • In-vitro diagnostic use.
  • The presence of bilirubin is an important finding in the evaluation of liver function.
  • Based on the diazotization reaction.

Specifications

Certifications/Compliance CLIA Waived
Content And Storage 15°C to 30°C
Clia Complexity Waived
Detectable Analytes Bilirubin
Sensitivity AimTabTM Bilirubin Tablets will detect as little as 0.05 to 0.1 mg/dL (0.8 to 1.7 μmol/L) bilirubin in urine.
Type Bilirubin Tablets
Includes 100 Tablets/bottle, absorbent mats, Instructions for use
Quantity 100/Bottle
Test Time 60 sec.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.