Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories UA Diptube Controls

Catalog No. 22111275
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-111-275 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-111-275 Supplier Germaine™ Laboratories Supplier No. 51142
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Controls used with Fisherbrand™ 10-SG Urine Reagent Strips

  • Conveniently packaged in diptubes for ease of use
  • 4 x 12mL (2 x 12mL negative, 2 x 12mL positive)
  • Store at 2° to 8°C

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Waived
Detectable Analytes Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Leukocyte
For Use With (Application) Urinalysis
Quantity 12 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.