Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories UA Liquid Controls

Catalog No. 22111276
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-111-276 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-111-276 Supplier Germaine™ Laboratories Supplier No. 51161
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid controls ready to use for Fisherbrand™ 10-SG Urine Reagent Strips

  • Conveniently packaged in dropper bottles for ease of use
  • 6 x 10mL (3 x 10mL negative, 3 x 10mL positive)
  • Store at 2° to 8°C

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Waived
For Use With (Application) Urinalysis
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.