Learn More
Electron Microscopy Sciences EMS Item German Autopsy Dissecting Knife, 11.75" OAL
SDP
Catalog No. 501931585
Encompass_Preferred
This dissection knife is lightweight and perfectly balanced with a hollow ground cutting edge. The handle has a smooth top edge for comfort and the blade is hardened to withstand continuous sharpening. The knife is made from premium grade stainless steel.
OAL: 11.75"
Width: 0.75"
Catalog No. 50-193-1585 Supplier Electron Microscopy Sciences Supplier No. 7205904
This dissection knife is lightweight and perfectly balanced with a hollow ground cutting edge. The handle has a smooth top edge for comfort and the blade is hardened to withstand continuous sharpening. The knife is made from premium grade stainless steel.
OAL: 11.75"
Width: 0.75"