Learn More
Electron Microscopy Sciences German Round Coverslips, 12 mm 1 OZ
SDP
Catalog No. 50949010
Encompass_Preferred
Cover slips for neutral cell cultures. Shown to be more effective with hard to attach cells. #1 Thickness (0.13 to 0.16 mm) and packed 1 oz (28g) per case.
Size: 12 mm
Catalog No. 50-949-010 Supplier Electron Microscopy Sciences Supplier No. 7219612
Cover slips for neutral cell cultures. Shown to be more effective with hard to attach cells. #1 Thickness (0.13 to 0.16 mm) and packed 1 oz (28g) per case.
Size: 12 mm