Learn More
Electron Microscopy Sciences German Round Coverslips, 25 mm 1 OZ
SDP
Catalog No. 50948982
Encompass_Preferred
Cover slips for neutral cell cultures. Shown to be more effective with hard to attach cells. #1 Thickness (0.13 to 0.16 mm) and packed 1 oz (28g) per case.
Size: 25 mm
Catalog No. 50-948-982 Supplier Electron Microscopy Sciences Supplier No. 7219625
Cover slips for neutral cell cultures. Shown to be more effective with hard to attach cells. #1 Thickness (0.13 to 0.16 mm) and packed 1 oz (28g) per case.
Size: 25 mm