Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Germanane

Catalog No. 9060261G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-602-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-602-61G Supplier Sigma Aldrich Supplier No. 9060261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Germanane (GeH) is a 2D graphane analog with a direct band gap of 1.6 eV and electron mobility of 18000 cm2V-1s-1. It is a hydrogen terminated germanium (Ge) which can be synthesized by the topotactic deintercalation of CaGe2 crystals. Its crystal structure contains hexagonal Ge lattice with hydrogen atoms that are covalently attached to every Ge atom.GeH is a van der Waals material that can be used in a variety of applications such as electronic, optoelectronic, sensing and energy conversion.

Specifications

CAS 13572-99-1
Molecular Formula GeH
Linear Formula GeH
MDL Number MFCD00011028
Quantity 1 g
Synonym Germanium hydride; Germanium monohydride; Germylidyne
Molecular Weight (g/mol) 73.65
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.