Learn More
Neuvitro Pre cleaned acid treated sterile German coverslip for cell culture, 12mm dia #1 90/pk
Catalog No. NC0217821
Encompass_Preferred
CE certified German glass coverslips for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, sterile. 12mm diameter round, #1 thickness, 90 pieces/pack. Ready to use directly in cell culture, or to add your own coating substrate. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 1 year when stored in chest freezer. User manual included.
Catalog No. NC0217821 Supplier Neuvitro Supplier No. GG12PRE
CE certified German glass coverslips for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, sterile. 12mm diameter round, #1 thickness, 90 pieces/pack. Ready to use directly in cell culture, or to add your own coating substrate. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 1 year when stored in chest freezer. User manual included.