Learn More
Simpore Inc 20 nm thick Ge over 20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 5
Catalog No. NC2818884
Encompass_Preferred
20 nm thick Ge over 20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 5
Catalog No. NC2818884 Supplier Simpore Inc Supplier No. GE20SN20MP2Q05
20 nm thick Ge over 20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 5