Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium Disulfide, MP Biomedicals

Catalog No. ICN204728 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204728 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204728 Supplier MP Biomedicals, Inc Supplier No. 0520472801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12025-34-2
Molecular Formula GeS2
Molecular Weight (g/mol) 136.75
MDL Number MFCD00016122
InChI Key YIZVROFXIVWAAZ-UHFFFAOYSA-N
Synonym germanium disulfide, germanium sulfide, bis sulfanylidene germane, dithioxogermane, germanic sulfide, germanium iv sulfide
PubChem CID 82816
IUPAC Name germanedithione
SMILES S=[Ge]=S

Specifications

Quantity 1 g
Formula Weight 136.72
Chemical Name or Material Germanium Disulfide
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.