Learn More
Strem, An Ascensus Company CAS# 12025-32-0. 2g. Germanium(II) sulfide (99.99+%-Ge) (contains 1-5% germanium metal) PURATREM. MFCD00135539
SDP
Catalog No. 5090115056
Encompass
CAS# 12025-32-0. 2g. Germanium(II) sulfide (99.99+%-Ge) (contains 1-5% germanium metal) PURATREM. MFCD00135539. Molecular Weight: 104.65. Molecular Formula: GeS. Color/form: black xtl. Strem# 93-3209. http://www.strem.com/catalog/v/93-3209/
Catalog No. 50-901-15056 Supplier Strem, An Ascensus Company Supplier No. 9332092G
CAS# 12025-32-0. 2g. Germanium(II) sulfide (99.99+%-Ge) (contains 1-5% germanium metal) PURATREM. MFCD00135539. Molecular Weight: 104.65. Molecular Formula: GeS. Color/form: black xtl. Strem# 93-3209. http://www.strem.com/catalog/v/93-3209/