Learn More
Strem, An Ascensus Company CAS# 1310-53-8. 5g. Germanium(IV) oxide (99.999%-Ge) PURATREM. MFCD00011030
SDP
Catalog No. 5090115062
Encompass
CAS# 1310-53-8. 5g. Germanium(IV) oxide (99.999%-Ge) PURATREM. MFCD00011030. Molecular Weight: 104.59. Molecular Formula: GeO2. Color/form: white pwdr., hexagonal (soluble) form Strem# 93-3221. http://www.strem.com/catalog/v/93-3221/
Catalog No. 50-901-15062 Supplier Strem, An Ascensus Company Supplier No. 9332215G
CAS# 1310-53-8. 5g. Germanium(IV) oxide (99.999%-Ge) PURATREM. MFCD00011030. Molecular Weight: 104.59. Molecular Formula: GeO2. Color/form: white pwdr., hexagonal (soluble) form Strem# 93-3221. http://www.strem.com/catalog/v/93-3221/