Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germanium(IV) oxide, 99.9999% (metals basis)

Catalog No. AA1051022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1051022 100 g
AA1051006 5 g
AA1051014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1051022 Supplier Thermo Scientific Chemicals Supplier No. 01051022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-53-8 | GeO2 | 104.628 g/mol

Germanium dioxide is used in algaculture.A mixture is used as an optical material for optical fibers and optical waveguides It is also used as a catalyst in production of polyethylene terephthalate resin and for production of other germanium compounds. It is used as a feedstock for production of some phosphors and semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1086°C
Color White
Physical Form Powder
Quantity 100 g
Odor Odorless
Assay Percent Range 99.9999% (Metals basis)
Density 6.239
Solubility Information Soluble in water (4.5g/L at 25°C),acid and alkali. Insoluble in Hydrofluoric acid and hydrochloric acid.
Formula Weight 104.59
Chemical Name or Material Germanium(IV) oxide
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 215-180-8
RTECSNumber LY5240000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.